Learn More
Julabo™ Spring Clamps for Shaking Water Bath
Brand:  Julabo™ 8970601
28.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10311742
  • 28.70€ / Each
Only null left
Estimated Shipment: 07-11-2025
Specifications
Holds 10 mL Erlenmeyer flask
SW22, SW23 shaking water bath
Spring Clamp