Learn More
Julabo™ Spring Clamps for Shaking Water Bath
Brand:  Julabo™ 8970609
63.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10115930
  • 64.60 EUR / Each
Only null left
Estimated Shipment: 20-10-2025
Specifications
Holds 1000 mL Erlenmeyer flask
SW22, SW23 shaking water bath
Spring Clamp