Learn More
SPEX™ SamplePrep Large-Capacity Freezer/Mill™ Vial Divider for Grinding Vial Set 6751
For use with SPEX SamplePrep™ Large-Capacity Freezer/Mill Cryogenic Grinder
Brand: SPEX™ SamplePrep 6807
327.88 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Makes one complete vial plus three spare center sections
- Sample capacity: 0.5 to 4.0mL
- Includes stainless-steel impactor, two end plugs
Specifications
Multi Vial Adapter | |
Ideal for high throughput applications | |
Spex SamplePrep Large Capacity Freezer/Mill | |
Microvial set |