Learn More
Sodium Sulfate Decahydrate, 97%, Reagent grade, Honeywell™
Product Code. 15602690 Shop All Honeywell Research Chemicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Reagent Grade, 97%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7727-73-3
Molecular Formula H20Na2O14S
Molecular Weight (g/mol) 322.186
MDL Number MFCD00149189
InChI Key RSIJVJUOQBWMIM-UHFFFAOYSA-L
Synonym sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o
PubChem CID 62649
ChEBI CHEBI:32586
IUPAC Name disodium;sulfate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]
Specifications
Melting Point 32.4°C
Color White
Quantity 2.5 kg
Linear Formula Na2SO4 · 10H2O
UN Number NONH for all modes of transport
Formula Weight 322.2g/mol
Percent Purity 97%
Grade Reagent
Packaging Poly Bottle
Appearance Crystals or granules
Density 1.46g/mL at 25°C
Chemical Name or Material Sodium Sulfate Decahydrate