Learn More
Sodium Hydroxide, 5M, Chem Lab™
128.00€ - 238.00€
Specifications
Color | Colorless |
---|---|
Concentration | 200g NaOH/l H2O = 5 N (±0.01/20°C) |
Density | 1.18g/mL |
Solubility Information | Soluble in water |
Odor | Odourless |
Physical Form | Liquid |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11903243
|
Chem Lab
CL05.1425.5000 |
5 L | 473mL |
128.00€
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11923253
|
Chem Lab
CL05.1435.9010 |
10 L | 10L |
238.00€
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Colorless | |
1.18g/mL | |
Odourless |
200g NaOH/l H2O = 5 N (±0.01/20°C) | |
Soluble in water | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium hydroxide 5 mol/l
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Sodium hydroxide, pellets a.r.