Learn More
Macherey-Nagel™ Sodium Citrate
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
Technique | TLC Micro Set A |
Quantity | 2.5 g |
Type | Sodium Citrate |
For Use With (Equipment) | For TLC Micro Set A |