Learn More
Biochrom Single Cell Holder
Single Cell Holder is an accessory for the Ultrospec 7500 and 2100 Pro Spectrophotometers with a 10 to 50 mm pathlength.
Brand: Biochrom 80-2106-07
480.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Single cell holder
- 10 to 50 mm pathlength
- Accessory for the Ultrospec 7500, 2100 Pro Spectrophotometer
Specifications
Ultrospec 7500, 2100 Pro, 3100, 3300, 4300, 5300, 6300 | |
1-position Cell Holder | |
Ultrospec 7500, 2100 Pro Spectrophotometer | |
Round |
Single Cell Holder | |
Reusable | |
10 to 50 mm |