Learn More
Thermo Scientific™ SilTite™ Ferrule Kits
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits.
Brand: Thermo Scientific™ 290MA195
Description
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits. The ferrule forms a strong, permanent, airtight seal around the capillary column.
- SilTite metal ferrules and nuts are made from the same material and have the same thermal expansion coefficient
- Base of the ferrule is flat and forms a perfect seal with the MS interface
- The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
- Unlike other ferrules, do not need re-tightening after installation
Specifications
Ferrule Kit | |
Thermo Scientific ISQ GC-MS Interface | |
2 SilTite Nuts, 10 Ferrules |
290 MA195 | |
SilTite Metal | |
0.32 mm |