Learn More
Epredia™ Signature Series™ Flex™ Dehydrating Reagent
Achieve brilliant cytoplasmic staining and complete dehydration with Epredia™ Signature Series Flex™ 100, 95, 80.
97.90€ - 144.00€
Product Code | Brand | Quantity | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Product Type | Price | Quantity & Availability | ||||
10550529
|
Epredia™
8101 |
1 gal. | Flex 100 Dehydrating Reagent |
144.00€
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10851560
|
Epredia™
8201 |
1 gal. | Flex 95 Dehydrating Reagent |
144.00€
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12638106
|
Epredia™
8301R |
1 gal. | Flex 80 Dehydrating Reagent | ||||||
Description
- Patented formulation of methyl and isopropyl alcohol minimizes over-dehydration in smaller biopsies
- Excellent for lipid extraction allowing for increased paraffin infiltration
- May be used as a solvent for special stains
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Flammable liquid Category 2
- Specific target organ toxicity Category 1
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H370-Causes damage to organs.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P271-Use only outdoors or in a well-ventilated area.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P235-Keep cool.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P363-Wash contaminated clothing before reuse.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : isopropyl alcohol, methyl alcohol