Learn More
Epredia™ Shandon™ Instrument Case
Epredia™ Shandon Instrument Case is available as a complete kit or case only.
3070.00€
Specifications
Product Type Instrument Case
View More Specs
Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15338278
View Documents
Epredia™
9996969
Case with instruments
3070.00€
1 set
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 18-12-2025
Add to basket
Description
Description
Case dimensions:
  • 12.75 x 9.5 x 2.5 in. (32.4 x 24.1 x 6.4cm)
Instruments Available in Varying Sizes and Styles:
  • Dissecting scissors
  • Iris scissors
  • Liston bone shears
  • Rib shears
  • Plain bone shears
  • Liston bone-cutting forceps
  • Councilman's chisel
  • Tissue thumb forcep
  • Broad point dressing thumb forceps
  • Cartilage thumb forceps
  • Halstead's hemostatic mosquito forceps
  • Dura strip forceps
  • Virchow skull breaker
  • Bone mallet
  • Post mortem hammer with hook
  • Grooved needle director
Specifications
Specifications
Instrument Case