Learn More
Epredia™ Embedding Cassettes in Tube Packs
Hold and identify tissue samples in processing, embedding and sectioning procedures with Embedding Cassettes in Tube Packs. Compatible for use with the Epredia™ PrintMate AS™ Cassette Printer.
Brand:  Epredia™ B1000732YW
 View more versions of this product
Product Code. 15145944
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • Can be used on all Epredia PrintMate automatic cassette labelers
  • Fit securely in microtome chuck adapters
  • Designed to accept metal lids (Order number 1001003, 25/pk., sold separately)
  • Made of acetal polymer with a pore style of round holes
  • Supplied in 10 tubes of 75 cassettes each
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Round Holes
750 Cassettes/Cs.
Yellow
Acetal Polymer