Learn More
Epredia™ Shandon™ Embedding Cassettes II in Tube Packs, aqua
Hold tissue specimens during processing and embedding with Epredia™ Shandon™ Embedding Cassettes II in Tube Packs.
178.00€ - 206.00€
Specifications
Pore Style | Slotted Pore |
---|---|
Writing Face Angle | 45° |
Product Type | Biopsy Cassette |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
12903755
|
Epredia™
B1000729AQ |
Aqua | N/A | ||||||
12654893
|
Epredia™
B1000729BL |
Blue | N/A | ||||||
12963755
|
Epredia™
B1000729GR |
Green | N/A | ||||||
12973755
|
Epredia™
B1000729GY |
Gray | N/A | ||||||
12983755
|
Epredia™
B1000729LI |
Lilac | N/A | ||||||
12993755
|
Epredia™
B1000729OR |
Orange | N/A | ||||||
12903765
|
Epredia™
B1000729PE |
Peach | N/A | ||||||
12913765
|
Epredia™
B1000729PK |
Pink | N/A | ||||||
12923765
|
Epredia™
B1000729TN |
Tan | N/A | ||||||
12513537
|
Epredia™
B1000729WH |
White | N/A | ||||||
12933765
|
Epredia™
B1000729YW |
Yellow | N/A | ||||||
Description
- Acetal polymer cassettes with writing face at 45° angle
- Slotted pore style
- Locking lid attached by two hinges
Ordering Information:
- Tube packs are supplied in 10 tubes of 75 cassettes each
Specifications
Slotted Pore | |
45° | |
Biopsy Cassette |