Learn More
Epredia™ Double Cytoslide™ Microscope Slides
Use Epredia™ Shandon™ Double Cytoslides™ with Epredia™ Shandon™ EZ Double Cytofunnel™ Chambers.
90.20€ - 94.30€
Specifications
Type Double Cytoslide
Color Plain
View More Specs
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
12006689
View Documents Promotion Details
Epredia™
5991054
Noncoated
90.20€
Pack of 100
Only null left
In Stock
Add to basket
12016689
View Documents Promotion Details
Epredia™
5991055
Coated
94.30€
Pack of 100
Only null left
In Stock
Edge
Add to basket
Description
Description
  • Specimen is displayed within a circle marked as 1 or 2
  • Number circles correspond with the Cytofunnel™ chamber
Ordering Alert:
  • Sold in boxes of 100 slides
Specifications
Specifications
Double Cytoslide
Plain