Learn More
Memmert™ Shaking Device for Waterbaths
Shaking device with support frame without gable cover & support basket (mandatory)
Brand:  Memmert™ WTB35SV3550
1493.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17180385
  • 1535.00€ / Each
Currently Unavailable: We have no information when this product will be available.
View alternative products
Description
Description
  • Requires gable cover
  • Voltage 230 V, 50/60 Hz
  • Order the support basket separately
  • Includes support frame, shaking frequency 35 - 160 strokes per minute (horizontal back/forth movement)
Specifications
Specifications
WTB35
Shaking Device
35-160 rpm