Learn More
Trajan™ Standard Plunger Protection Syringes
Click to view available options
Volume (Metric):
5 μL
Unit Size:
Each
Description
- MicroVolume syringe SuperfleX plunger
- Bevel tip
Specifications
Specifications
Needle Gauge | 26 G |
Diameter (English) Outer Needle | 0.018 in. |
Diameter (Metric) Outer Needle | 0.47 mm |
Length (English) Needle | 1.96 in. |
Product Type | Microvolume Syringe |
Model | 5FX |
Needle Point Number | 2 |
Quantity | Ea. |
Volume (Metric) | 5 μL |
Needle Point Style | Bevel |