Learn More
SGE™ ConnecTite™ Liners for Shimadzu™ GC
Designed for use with Shimadzu 2010™ GC
391.00€ - 498.00€
Specifications
Length (Metric) | 9.5 cm |
---|---|
Diameter (Metric) Inner | 0.34 cm |
Diameter (Metric) Outer | 0.5 cm |
For Use With (Equipment) | Shimadzu 2010 |
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
15219023
|
SGE™
092331 |
Connectite Bottom Hole Liner | 5/Pk. |
498.00€
Pack of 5 |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||
15279013
|
SGE™
092329 |
Connectite Standard Liner | 5/Pk. |
391.00€
Pack of 5 |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||
15299013
|
SGE™
092330 |
Connectite Top Hole Liner | 5/Pk. |
498.00€
Pack of 5 |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||
Description
- Range covers all applications and all popular instrument makes/ models
- Range covers vaporizing and cool on-column injection techniques
- Inlet liners ensure complete transfer of sample from syringe to the column without loss
- 5mm OD, 3.4mm ID; 95mm length
Specifications
9.5 cm | |
0.34 cm | |
0.5 cm | |
Shimadzu 2010 |