Learn More
Cytiva Sentino™ Microbiology Pump Replacement Adapter
Click to view available options
:
Stainless Steel
Description
- For vacuum filtration of liquids
- For the analysis of aqueous samples for microbiological and particulate contamination using the Membrane Filter (MF) Technique
- For research use only
Specifications
Specifications
For Use With (Equipment) | MicroFunnel™ Filter Funnel |
Material | Stainless Steel (316L) |
Sterility | Non-sterile |