Learn More
IKA™ Scale with Stand
Scale with a stand that provides increased comfort and safety for laboratory applications.
Brand:  IKA™ 0025004318
Product Code. 16238641
  • 981.00€ / Each
Only null left
In Stock
Add to basket
Description
Description
  • A stand with an integrated scale and data interface
  • Sample weight of up to 6 kg - The intelligent stand scale has a weighing range of up to 6 kg with a resolution of 1 g
  • Particularly suitable for the direct and controlled addition of sample material while stirring
  • Clear display - Working with the clear and modern LCD display is very userfriendly
  • All information, whether measurement values, calibration steps or status messages, is shown clearly
  • Stable set-up plate - The solid die cast zinc housing provides stability and robustness
  • Bluetooth connection - The weight of the sample measured can directly be transferred to a suitable device (e.g. IKA Mini-/Microstar series) and shown on its display
TRUSTED_SUSTAINABILITY
Specifications
Specifications
213 x 570 x 391 mm
1