Learn More
Masterflex™ Anti-Siphon Valves
Connect these valves to bio-bags or sampling tubes. Cole-Parmer™ Anti-Siphon Valves are molded onto the tubing for a seamless transition.
Brand:  Masterflex™ 30505-91
Product Code. 13513010
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
Valves feature low priming volume, ease of removing air bubbles during priming and operate in any spatial orientation. Manufactured DEHP-and latex-free silicone diaphragms. Materials meet USP CLass VI and ISO 10993 specifications. Each assembly comes packaged individually and can be sterilized..
Specifications
Specifications
SAN
10 Pack
Luer Fitting