Learn More
Thermo Scientific™ Sample Sensor for Thermo Scientific™ Heratherm™ Advanced Protocol Security Incubators
Measures exact temperature
Brand: Thermo Scientific™ 50127768
306.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For connection to all Advanced Protocol Security incubators
- Measures exact sample temperature
- Temperature shown on display as plugged in
- Cable length: 86.6 in.
Specifications
| Advanced Protocol Security Incubators | |
| Sensor |