Learn More
Kimble Chase™ Round Bottom Borosilicate Glass Culture Tube
554.00€ - 559.00€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11888482
|
Kimble Chase™
73770-20125 |
25 mL |
554.00€
Pack of 500 |
Estimated Shipment: 15-07-2025
Log in to see stock. |
|||||
11843133
|
Kimble Chase™
73770-20150 |
30 mL |
559.00€
Pack of 500 |
Estimated Shipment: 02-09-2025
Log in to see stock. |
|||||
Description
Specifications
Single-use | |
500Pack | |
Culture Tube |