Learn More
Eppendorf™ Rotor Adapters for Rotor S-4x1000
Shop All Eppendorf Products
Click to view available options
Capacity:
25 mL, 50 mL
5.0 mL and 15 mL
5.5 to 12 mL
For Use With (Equipment):
For Rotor S-4x1000 high-capacity buckets
For Rotor S-4x1000 plate/tube buckets
Unit Size:
Pack of 2
Description
Adapters for Rotors. Available for use with:
- 5.5 to 12mL, set of 2 pcs.
- 50mL, set of 2 pcs.
- Eppendorf Tubes 5.0mL and 15mL, set of 2 pcs.
Specifications
Specifications
Capacity | 5.0 mL and 15 mL |
Max. Tube Diameter | 17mm |
For Use With (Equipment) | For Rotor S-4x1000 plate/tube buckets |
Description | Conical bottom |
Vessels per Adapter | 16 |
Max. RCF | 3076 x g |
No. of Places | 16/64 |
Tube Size | Eppendorf Tubes 5.0mL and 15mL conical |