Learn More
Thermo Scientific Chemicals Ringer's Solution, standard, unbuffered
97.79€ - 149.75€
Specifications
Chemical Name or Material | Ringer's Solution |
---|---|
Physical Form | Liquid |
Synonym | dihydrogen oxide, dihydrogen monoxide |
Packaging | Plastic bottle |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15408759
|
Thermo Scientific Chemicals
J67524.AP |
500 mL |
97.79€
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15418759
|
Thermo Scientific Chemicals
J67524.K2 |
1 L |
149.75€
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ringer's Solution | |
Liquid | |
dihydrogen oxide, dihydrogen monoxide | |
Plastic bottle |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures