Learn More
BINDER™ Access Port with Silicone Plug, Right Access
BINDER™ Right Access Port is an accessory for use on the right of a variety of chambers and incubators.
Brand:  BINDER™ 8012-1333
934.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15902982
  • 1030.00€ / Each
Only null left
Estimated Shipment: 07-11-2025
Description
  • For use on right
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Specifications
Access port with silicone plug - 80 mm, right
Silicone
Silicone plug
BINDER Chambers Series MKT
Access Port
80 mm