Learn More
BINDER™ Access Port with Silicone Plug, Right Access
BINDER™ Right Access Port is an accessory for use on the right of a variety of chambers and incubators.
Brand: BINDER™ 8012-1349
990.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use on right
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Access port with silicone plug - 125 mm, right | |
Silicone | |
Silicone plug |
BINDER Chambers Series MKF | |
Access Port | |
125 mm |