Learn More
Epredia™ Papanicolaou OG-6 and Modified OG Stains
Shop All Epredia Products
Click to view available options
Quantity:
1 gal.
Type:
OG-6 Papanicolaou Stain
Unit Size:
3.79 L
Description
- OG-6 stains the keratinized areas of the cell
- Modified OG is a Gill modification that minimizes precipitation and reduces staining time
- Use OG-6 with EA-36 or EA-50 when staining gynecological specimens
- Use modified OG with EA-65 for staining non-gynecological specimens
Specifications
Specifications
Type | OG-6 Papanicolaou Stain |
Quantity | 1 gal. |
Product Content Correction
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion