Learn More
Grant Instruments™ Polycarbonate Transparent Lid, Gabled
Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
92.22 EUR - 162.00 EUR
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11496417
|
Grant Instruments™
AQL12 |
Model SAP12, JBN12, JBA12 and SBB Aqua 12 Plus SUB Aqua Plus water bath |
146.00 EUR
Each |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
11406427
|
Grant Instruments™
AQL26 |
26L Tanks |
162.00 EUR
Each |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
11486417
|
Grant Instruments™
AQL5 |
5L Tanks. Fits the Grant JBA5 bath, and would be a valid replacement lid for the JBN5. |
119.00 EUR
Each |
Estimated Shipment: 03-10-2025
Log in to see stock. |
|||||
11476417
|
Grant Instruments™
AQL2 |
2L Tanks | |||||||
Description
Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
- Polycarbonate construction
- Clear, gabled lid allows condensation buildup to drip away from immersed vessels.
- For use with Sub Aqua Pro Series Unstirred Water Baths
Specifications
Gabled | |
Polycarbonate |