Learn More
Olympus™ Reflected Light Brightfield Mirror Unit, U-MBF3
Use this cube-shaped mirror unit for brightfield illumination. Olympus™ Reflected Light Brightfield Mirror Unit, U-MBF3 is a narrow-style for the 6-position turret on the BX and IX series of microscopes.
Brand: Olympus™ K10017462
This item is not returnable.
View return policy
Description
- Half-mirror for reflected light brightfield illumination.
- Includes filter mirror set plus blank cube.
- Narrow style for the 6-position turret on the Olympus BX2 and IX2 series microscopes.
- Also retrofits the older 4-position turret on the BX and IX series microscopes.
Specifications
Reflected Light, Darkfield Mirror Unit | |
BX2, IX2, BX, IX microscopes |