Learn More
Thermo Scientific™ Racks and Inserts for Refrigerated and Heated Bath Circulators
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Brand: Thermo Scientific™ 1600086
187.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Specifications
Rack Insert; Includes top and bottom panels | |
Rack Insert | |
Stainless Steel |
Bath Types S13, S12T, S15 | |
8 25 mm Tubes |