Learn More
Thermo Scientific™ Racks and Inserts for Refrigerated and Heated Bath Circulators
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Brand: Thermo Scientific™ 1600003
168.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Specifications
Rack Insert; Includes top and bottom panels | |
Rack Insert | |
Stainless Steel |
Bath Types A5B, A10B, A24B, S49, S19T, S14P, S21P | |
100 10 mm Tubes |