Learn More
Quickfit™ Tubing Adapter with Side-arm
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
26.05€ - 177.00€
Product Code | Brand | Cone Size | Standard Taper Joint No. | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Standard Taper Joint No. | Price | Quantity & Availability | ||||
12028371
|
Quickfit
MF17/1/SC |
14/23 | 14/23 |
26.05€
Each |
Estimated Shipment: 21-10-2025
Log in to see stock. |
||||
10440485
|
Quickfit
MF18/2/SC |
19/26 | 19/26 |
34.65€
Each |
Estimated Shipment: 03-12-2025
Log in to see stock. |
||||
12048371
|
Quickfit
MF18/3/SC |
24/29 | 19/26 |
32.45€
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
12058371
|
Quickfit
MF18/33/SC |
24/29 | 24/29 |
34.65€
Each |
Estimated Shipment: 23-09-2025
Log in to see stock. |
||||
12455316
|
Quickfit
MF18/44/SC |
29/32 | 29/32 |
32.60€
Each |
Estimated Shipment: 21-10-2025
Log in to see stock. |
||||
12465316
|
Quickfit
MF18/5/SC |
34/35 | 19/26 |
32.60€
Each |
Estimated Shipment: 09-12-2025
Log in to see stock. |
||||
15507940
|
Quickfit
MF18/4/SC |
29/32 | 19/26 |
177.00€
Pack of 5 |
Estimated Shipment: 09-12-2025
Log in to see stock. |
||||
15587930
|
Quickfit
MF17/2/SC |
19/26 | 14/23 |
145.00€
Pack of 5 |
Estimated Shipment: 09-12-2025
Log in to see stock. |
||||
15597930
|
Quickfit
MF17/3/SC |
NS 24/29 | NS 24/29 |
148.00€
Pack of 5 |
Estimated Shipment: 21-10-2025
Log in to see stock. |
||||
Description
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
- With sidearm for the attachment of rubber hose etc
- Detachable plastic screwthread connector allows flexible tubing to be fitted easily and safely