Learn More
BRAND™ PTFE Ground Sockets
Constructed of PTFE. BRAND™ Ground Sockets with greaseless seal will not contaminate sample. Outstanding chemical resistance at operating temperatures from -200 to +260°C. Grip collar and sealing rings provide an oustanding tight seal and low leakage rate. Sturdy design allows permanent use.
Brand: BRAND™ 51467
26.40 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Constructed of PTFE.
- Greaseless seal will not contaminate sample.
- Outstanding chemical resistance at operating temperatures from -200 to +260°C.
- Sturdy design for permanent use.
- With grip collar and sealing rings for excellent tightness.
- Low leakage rate: Q < 1 · 10-4 mbar · l · s-1.
Specifications
Grip collar | |
PTFE | |
Ground socket | |
NS 29/32 |