Learn More
Kartell™ Lining for Glass Tapers
Constructed of PTFE for high temperature and chemical resistance. Kartell™ Lining for Glass Tapers are specially designed to prevent machined glass joints from binding.
Brand: Kartell™ 0103900
22.64 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Designed to prevent machined glass joints from binding
- Conforms to DIN 12242
- Suitable for foodstuff
Specifications
DIN 12242 | |
16 mm | |
Prevent glass joints from binding | |
19/26 Cone Size |
PTFE | |
Glass Joint | |
10/Pk. | |
Ground Joint Sleeve Lining |