Learn More
Semadeni™ PS Boxes
2.00€ - 4.00€
Specifications
Closure Type | Lid |
---|---|
Color | Transparent |
Material | Polystyrene |
Shape | Rectangular |
For Use With (Application) | Packaging, Storage |
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | |||||
11802310
|
Semadeni™
1729 |
94 x 68 x 29 mm |
2.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11812320
|
Semadeni™
1733 |
105 x 87 x 50 mm | N/A | ||||||
Specifications
Lid | |
Polystyrene | |
Packaging, Storage | |
Storage Box |
Transparent | |
Rectangular | |
Tightly Closing Lid | |
1 Each |