Learn More
DuPont™ Proshield™ 4 Practik
Proshield™ 4 Practik suits are the ideal work suits for general maintenance in industrial manufacturing and other harmless environments.
46.11€ - 184.44€
Specifications
Gender | Unisex |
---|---|
Material | Polyethylene |
Color | White |
Disposable | Single-use |
Cuff Style | Elastic |
Description
- Micro-perforated non-woven polyethylene fabric allows unrestricted passage of air and water vapour
- Certified according to Regulation (EU) 2016/425
- Chemical protective clothing, Category I
- Antistatic treatment (EN 1149-5) on inside for comfort
- Versatile protective garment for non-hazardous substances
Specifications
Unisex | |
White | |
Elastic | |
Zipper | |
Elastic |
Polyethylene | |
Single-use | |
Hooded | |
Non-sterile |