Learn More
uvex Profastrong NF33 Chemical Protection Gloves
Sensitive NBR chemical protection gloves feature a flocked cotton lining for comfort and provide a high degree of mechanical abrasion resistance.
12.65€
Specifications
Color | Green |
---|---|
Latex Free | Yes |
Material | NBR |
Product Type | Chemical-resistant Gloves |
EN388 Standard | 4101X |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15386922
|
uvex
6012202 |
8 |
12.65€
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
|||||
15396922
|
uvex
6012203 |
9 |
12.65€
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
15366922
|
uvex
6012204 |
10 |
12.65€
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
|||||
15376922
|
uvex
6012201 |
7 |
12.65€
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
|||||
Description
- Sensitive NBR chemical protection gloves with flocked cotton lining for comfort
- Embossed structure on the palm enhances grip capabilities in damp and wet areas
- Nitrile rubber is resistant to oils, grease, acids, and alkalis
- Gloves provide a tactile feel, secure fit, and mechanical abrasion resistance
- Ideal for use in various industries including automotive, chemical, and printing
Specifications
Green | |
NBR | |
4101X | |
AJKLOT | |
Gauntlet | |
33 cm |
Yes | |
Chemical-resistant Gloves | |
Type A | |
Bacteria, Fungi WITH Virus Protection | |
Cotton |