Learn More
Miele Professional Process Documentation
APH 110 Net500 Segosoft network converter for connection of machines with serial interface for a network.
Brand:  Miele Professional 9687440
1150.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 30130359
  • 1340.00€ / 1 unit
Only null left
Estimated Shipment: 28-08-2025
Description
  • Converts serial data (RS 232) to network protocol (TCP/IP)
  • Galvanic separation of all connections including surge protection
  • Power over Ethernet (PoE) enabled
  • For use with Segosoft Miele Edition
  • Scope of delivery: plug-in power unit (1.4 m)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Software