Learn More
Bel-Art™ SP Scienceware™ Glove Box Port Covers
Use these glove box port covers to seal off any glove ports to which gloves are not attached
Brand: Bel-Art™ SP Scienceware™ H50025-0008
311.65 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Simply press the covers into the port opening until the flange on the cover meets the glove port
- One-piece construction with a neoprene gasket for a snug fit
- Fit glove ports with a 20.3cm (8 in.) O.D.
Specifications
Glove Box Port Cover | |
Black | |
One-Piece | |
8 in. Glove Box Ports |