Learn More
Mutexil™ Visitor Protective Lab Coats
Use Mutexil™ Visitor Protective Lab Coats for visitors' safety and protection when visiting a facility that merits this required protective clothing.
Brand: Mutexil™ 4574739-XXL
81.40 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
PP non woven labcoat with press-stud fastening and pockets- Disposable
- PP, 40g/m2
- For maintenance, production site visits
- For non hazardous environments
Specifications
Lab Coat | |
White | |
None | |
Non-woven |
2X-Large | |
Snaps | |
Individually bagged |