Learn More
Mutexil™ Polyethylene Embossed Apron
Click to view available options
Thickness:
20 μ
25 μ
40 μ
70 μ
Specifications
Specifications
Type | Embossed Apron |
Material | Polyethylene |
Color | White |
Thickness | 40 μ |
Length (Metric) | 120 cm |
Width (Metric) | 70 cm |