Learn More
Thermo Scientific™ Plate Count Agar (Dehydrated)
Determine the plate counts of micro-organisms in food, dairy samples, waste water and other material of sanitary importance with this medium.
125.00€ - 1035.00€
Specifications
Certifications/Compliance | Industrial Reference: NMKL. |
---|---|
Description | Plate Count Agar |
Form | Powder |
Product Type | Agar |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10331433
|
Thermo Scientific™
CM0325B |
500 g |
125.00€
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11429688
|
Thermo Scientific™
CM0325R |
2.5kg |
1035.00€
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Determine the plate counts of microorganisms in food, dairy samples, waste water and other material of sanitary importance with Thermo Scientific™ Oxoid™ Plate Count Agar (Dehydrated). Plate Count Agar was developed by Buchbinder et al1 which is recommended by APHA2, AOAC3 and PHLS4. The quality control of Plate Count Agar (APHA) includes testing in accordance with ISO 11133:2014.5
Plate Count Agar is equivalent to the medium recommended by the APHA1, PHLS 2 and ISO3,4 for the plate count of micro-organisms in food.
Specifications
Industrial Reference: NMKL. | |
Plate Count Agar | |
Powder | |
Agar |
For Laboratory Use Only