Learn More
SymLine™ Pipe Connection Pieces
These pipe connection pieces provide connection to the central collection point. Place the collecting container or canister where you want it: the pipe system can be exactly configured to suit your needs!
134.00€ - 283.00€
Specifications
Closure Color | Black |
---|---|
Liner Material | HDPE-EC |
For Use With (Application) | Connecting Pipes with 32 mm OD |
For Use With (Equipment) | Tube System with SymLine™ FlexTube and FlexTubePro, Connectors, Distributors and Shut-off |
Diameter (Metric) Inner | 32 mm |
Product Code | Brand | Closure Size | Includes | Design | Length (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Includes | Design | Length (Metric) | Price | Quantity & Availability | ||
15514515
|
SymLine™
106427 |
32 mm (f) | PTFE-EL Sealing Ring, Knurled Nut | Straight | 118 mm |
156.00€
Each |
Estimated Shipment: 12-09-2025
Log in to see stock. |
||
10233090
|
SymLine™
106430 |
32 mm (f) to 32 mm (m) | PTFE-EL Sealing Ring, Knurled Nut | Angled | 150 mm |
134.00€
Each |
Estimated Shipment: 11-09-2025
Log in to see stock. |
||
10754432
|
SymLine™
106440 |
32 mm (f) to 32 mm (m) | PTFE-EL Sealing Ring, Knurled Nut | Angled | 500 mm |
149.00€
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
||
14746901
|
SymLine™
106421 |
32 mm (f) to 32 mm (m) | PTFE-EL Sealing Ring, Knurled Nut | Curved | 90 mm |
143.00€
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
||
15367439
|
SymLine™
106456 |
32 mm (f) to GL25 (m) | PTFE-EL Sealing Ring, Knurled Nut | Curved | 90 mm |
171.00€
Each |
Estimated Shipment: 25-09-2025
Log in to see stock. |
||
15813432
|
SymLine™
106458 |
2 x 32 mm (f), 1 x GL25 (m) | PTFE-EL Sealing Ring, Knurled Nut | T-piece | 118 mm |
281.00€
Each |
Estimated Shipment: 25-09-2025
Log in to see stock. |
||
13528840
|
SymLine™
106426 |
3 x 32 mm (f) | PTFE-EL Sealing Ring, Knurled Nut | T-piece | 118 mm |
267.00€
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
||
15844435
|
SymLine™
106476 |
3 x GL25 (m) | Manifold with 2 Fixation Holes | Manifold | 68.25 mm |
167.00€
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
||
15803432
|
SymLine™
106457 |
32 mm (f) to GL25 (m) | PTFE-EL Sealing Ring, Knurled Nut | Elbow | 75.5 mm |
194.00€
Each |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||
18686478
|
SymLine™
106732 |
32 mm (f) | Pipe Extension | Straight | 1200 mm |
283.00€
1 sheet |
Estimated Shipment: 15-09-2025
Log in to see stock. |
||
Description
Specifications
Black | |
Connecting Pipes with 32 mm OD | |
32 mm |
HDPE-EC | |
Tube System with SymLine™ FlexTube and FlexTubePro, Connectors, Distributors and Shut-off | |
Connection for Pipe System |