Learn More
Ohaus™ Pioneer™ PX 3202/E Precision Balance
Product Code. 15875337 Shop All Ohaus Products
Click to view available options
:
Capacity: 4200 g
The Pioneer PX combines essential weighing functionality with competitive performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings.
  • Competitive Performance at an Economical Price - The PX offers high accuracy and repeatability for essential weighing applications in laboratory, industrial and educational settings at an economical pricepoint.
  • Durably Constructed for Long-Term Use - Featuring a cast metal lower housing, sub-pan and stainless steel weighing pan, the PX is durably constructed for versatile, long-term use.
  • Designed for Simple, Intelligent Operation - The PX features a second line display for additional information or guidance, a static removal bar for convenient grounding (models with draftshield), and USB and RS232 connectivity for easy communication.
TRUSTED_SUSTAINABILITY
Specifications
Calibration Not Applicable
Certifications/Compliance CE
Connection Type RS232 & USB
Description Electronic Balance, PX3202/E
Capacity 3200 g
Display Type Backlit LCD
Readability 0.010000 g
Languages English (US), French, German, Italian, Spanish (MX)
Linearity 0.020000 g
Material ABS Plastic, Cast Metal, Stainless Steel, Glass
Model PX3202/E
Repeatability 0.010000 g
Stabilization Time 1.000 sec.
Weighing Modes Basic Weighing, Parts Counting, Percent Weighing, Animal Weighing, Density Determination
Dimensions (D x W x H) 321 x 209 x 98 mm
Dimensions (L x W x H) 321.00 x 209.00 x 98.000 mm
Length (Metric) 321.00 mm
Width (Metric) 209.00 mm
Height (Metric) 98.000 mm
Depth (Metric) 321.00 mm
Diameter (Metric) Outer Pan 180 mm
Weight (Metric) 3.5 kg
Weight (Metric) Shipping 5 kg
Electrical Requirements 100/240 V ∼200 mA 50/60 Hz 12/18 VA