Learn More
Cole-Parmer™ PFA Threaded Plug
Threaded fittings offer more permanent installation
36.05€ - 63.90€
Specifications
Type | Plug |
---|---|
Color | Clear |
Port Type | Male NPT Fitting |
Sterilization Method | Ethylene Oxide (ETO) Only |
Overall Length (English) | 27/32 in. |
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
11775758
|
Cole-Parmer™
06546-40 |
1/2 in. |
63.90€
Each |
Estimated Shipment: 27-10-2025
Log in to see stock. |
|||||
11755758
|
Cole-Parmer™
06546-39 |
3/8 in. |
36.05€
Each |
Estimated Shipment: 27-10-2025
Log in to see stock. |
|||||
Description
- Chemically-inert PFA handles most aggressive chemicals
- Threaded reducing bushing
- Choose the size and material that meets your application needs
- Use with our rigid tubing for your high-pressure applications
Specifications
Plug | |
Male NPT Fitting | |
27/32 in. |
Clear | |
Ethylene Oxide (ETO) Only | |
PFA |