Learn More
3M™ PELTOR™ X3 Series Ear Muffs
PELTOR™ X3 Series Ear Muffs in both over-the-head and hard hat-attached styles feature high attenuation ideal for moderate to high level noise environments.
52.00€
Specifications
Color | Black, Red |
---|---|
ANSI S3.1974 | Yes |
CSA Z94.2 | Class AL |
Cushion Color | Black |
Pairs per Package | 1 Ea. |
Product Code | Brand | Model | Dielectric | Style | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Dielectric | Style | Price | Quantity & Availability | |||
13409928
|
3M™
7000103991 |
X3A | Yes | Over-the-head | |||||
Description
- Appropriate for moderate to high noise exposure
- Twin headband design helps reduce heat buildup with good fit and balance
- Ear cups ensure a customized fit and comfort for individual users
- Convenient storage
- Compatibility with hard hats (models X3P3E and X3P5E)
- Dielectric (excludes model X3P3E)
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Black, Red | |
Class AL | |
1 Ea. |
Yes | |
Black | |
X3 Series |