Learn More
Thermo Scientific™ Oxoid™ Vegetable Peptone Broth
Thermo Scientific™ Oxoid Vegetable Peptone Broth is a meat-free general-purpose peptone.
112.00€
Specifications
Content And Storage | Dehydrated Powder, Ambient & Dry Storage |
---|---|
Form | Dehydrated Powder |
Product Type | Peptone |
For Use With (Application) | Culture media or fermentation processes |
Protein Source | Pea / Yeast |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10066842
|
Thermo Scientific™
VG0101B |
500 g |
112.00€
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific™ Oxoid Vegetable Peptone Broth is a highly nutritious, general-purpose peptone.
An exceptional meat-free alternative to Typtone Soya Broth, it delivers equivalent performance in culture media or as a fermentation base.
With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance, and scale to ensure successful microbial vaccine production.
Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.
Supports quality media design:
- Balanced nutrient composition
- Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
- Highly soluble
- BSE and mycoplasma free
Multi-method suitability:
- Batch culture
- Fed-batch culture
- Continuous culture
- Cell culture
Specifications
Dehydrated Powder, Ambient & Dry Storage | |
Peptone | |
Pea / Yeast |
Dehydrated Powder | |
Culture media or fermentation processes |
Safety and Handling
missing translation for 'shelfLife' : 720 Days from Manufacture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.