Learn More
Thermo Scientific™ Oxoid™ AnaeroJar™ Lid
Oxoid AnaeroJar Lid is for use with AnaeroJar 2.5L, Part No. AG0025A.
Brand: Thermo Scientific™ AG0027A
136.68 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Oxoid AnaeroJar Lid is for use with AnaeroJar 2.5L, Part No. AG0025A.
Specifications
AnaeroJar Lid | |
Oval Lid | |
Thermo Scientific Oxoid | |
1 Ea. |