Learn More
Thermo Scientific™ Owl™ Accessories for CST Multiple Gradient Gel Casters
Choose between stainless-steel clamps or binder clamps from Thermo Scientific™ Owl™ Accessories for CST Multiple Gradient Gel Casters.
57.50€ - 79.10€
Specifications
For Use With (Equipment) | Owl CST System |
---|---|
Product Line | Owl |
Quantity | 12 Pk. |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
12444587
|
Thermo Scientific™
CL-12 |
Binder Clamp |
57.50€
Each |
Estimated Shipment: 09-10-2025
Log in to see stock. |
|||||
12454587
|
Thermo Scientific™
CL-12S |
Clamp |
79.10€
Each |
Estimated Shipment: 09-10-2025
Log in to see stock. |
|||||
Description
Choose between stainless-steel clamps or binder clamps from Thermo Scientific™ Owl™ Accessories for CST Multiple Gradient Gel Casters. Packaged in sets of 12, these clamps are for use with Owl CST Multiple Gradient Gel Casters.
Specifications
Owl CST System | |
Owl | |
12 Pk. |