Learn More
Ohaus™ OIML F1 Weight Set, NVLAP Certificate
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML F1 Tolerances
417.00€ - 2720.00€
Specifications
Certifications/Compliance | NVLAP Accredited Certificate |
---|---|
Includes | Protective Case |
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
16622482
|
Ohaus™
80780435 |
500 mg to 1 mg |
417.00€
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
16632482
|
Ohaus™
80780437 |
50 g to 1 mg |
873.00€
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
16642482
|
Ohaus™
80780438 |
50 g to 1 g |
498.00€
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
16652482
|
Ohaus™
80780439 |
100 g to 1 mg |
944.00€
Each |
In Stock
Log in to see stock. |
|||||
16662482
|
Ohaus™
80780440 |
100 g to 1 g |
575.00€
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
16672482
|
Ohaus™
80780441 |
2 x 200 g to 1 mg |
1130.00€
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
16682482
|
Ohaus™
80780442 |
2 x 200 g to 1 g |
728.00€
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
16692482
|
Ohaus™
80780445 |
500 g to 1 mg |
1235.00€
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
16602492
|
Ohaus™
80780446 |
500 g to 1 g |
861.00€
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
16612492
|
Ohaus™
80780447 |
1 kg to 1 mg |
1685.00€
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
16622492
|
Ohaus™
80780448 |
1 kg to 1 g |
1510.00€
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
16632492
|
Ohaus™
80780449 |
2 x 2 kg to 1 mg |
2270.00€
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
16642492
|
Ohaus™
80780450 |
2 x 2 kg to 1 g |
2175.00€
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
16652492
|
Ohaus™
80780453 |
5 kg to 1 mg |
2720.00€
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
16662492
|
Ohaus™
80780454 |
5 kg to 1 g |
2585.00€
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
Description
- Weight Sets in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
- NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
NVLAP Accredited Certificate | |
Protective Case |