Learn More
Thermo Scientific™ O'RangeRuler DNA Ladders, ready-to-use
Thermo Scientific O'RangeRuler DNA Ladders is a step ladder designed for precise sizing of DNA fragments in agarose or non-denaturing polyacrylamide gels.
129.00€ - 183.00€
Specifications
Concentration | 6X |
---|---|
Gel Type | Agarose |
Label or Dye | Xylene Cyanol FF, Orange G |
Product Type | DNA Ladder |
Ready to Load | Yes |
Product Code | Brand | Size Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size Range | Price | Quantity & Availability | |||||
10839020
![]() |
Thermo Scientific™
SM1303 |
10 to 100 bp |
183.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10765271
![]() |
Thermo Scientific™
SM1313 |
10 to 100 bp |
168.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10598170
![]() |
Thermo Scientific™
SM1323 |
10 to 100 bp |
152.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10181120
![]() |
Thermo Scientific™
SM0613 |
50 to 1000 bp |
144.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10191120
![]() |
Thermo Scientific™
SM0623 |
100 to 500 bp |
135.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10840041
![]() |
Thermo Scientific™
SM0653 |
100 to 6000 bp |
131.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10394810
![]() |
Thermo Scientific™
SM0633 |
200 to 3000 bp |
129.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10810231
![]() |
Thermo Scientific™
SM0643 |
500 to 6000 bp |
133.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific O'RangeRuler DNA Ladders are designed for precise sizing of PCR products and other double-stranded DNA fragments in agarose or non-denaturing polyacrylamide gels.
O'RangeRuler series is characterized by its 'step ladder' design. The chromatography-purified DNA fragments are precisely separated by a consistent increment (e.g., 5 bp, 10 bp, 20 bp, 50 bp, 100 bp, 200 bp, or 500 bp). The design enables accurate and precise sizing of DNA fragments. Not recommended for DNA quantification.
All O'RangeRuler DNA Ladders are supplied with 6X Orange DNA Loading Dye for sample DNA. Orange G dye can be used to monitor DNA migration in agarose or polyacrylamide gels and migrates at 50 bp in a 1% agarose gel.
Highlights
- Step ladders available in a wide range of increments from 5-to-500 bp
- Evenly spaced reference bands
- Sharp bands, easy-to-remember fragment sizes
- Ready-to-use—premixed with 6X Orange Loading Dye for direct loading
- Stable at room temperature for up to 24 months
Sustainability features
Ready-to-use O'RangeRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.
Specifications
6X | |
Xylene Cyanol FF, Orange G | |
Yes | |
Sustainable packaging |
Agarose | |
DNA Ladder | |
Agarose Gels | |
O'GeneRuler, O’RangeRuler |
Safety and Handling
missing translation for 'shelfLife' : 24 months when stored at room temperature.